Learn More
Thermo Scientific™ Adjustable Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Place these adjustable rack holders at any angle to increase sample aeration. They offer exceptional flexibility and incredible convenience for 10 to 30 mm diameter tubes.
6313.65 NOK - 6701.64 NOK
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
11881331
|
Thermo Scientific™
600076 |
Adjustable test tube rack | 10-13 mm (3-7mL) tubes |
6701.64 NOK
Each |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
11881281
|
Thermo Scientific™
600077 |
Adjustable angle test tube rack holder with test tube rack | MaxQ 8000 Shaker |
6313.65 NOK
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11891331
|
Thermo Scientific™
600078 |
Adjustable test tube rack | 21-25 mm (30mL) tubes |
6466.50 NOK
Each |
Estimated Shipment: 07-11-2025
Log in to see stock. |
|||||
11769438
|
Thermo Scientific™
600079 |
Adjustable test tube rack | 26-30 mm (50mL) tubes |
6454.74 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
Place these adjustable rack holders at any angle to increase sample aeration. Thermo Scientific™ Adjustable Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide added convenience and will accommodate test tube diameters from 10 to 30 mm. These rack holders deliver exceptional flexibility — the right choice for shaking tubes.
- Can be used only with their corresponding rack size
- Includes one test tube rack
Specifications
Tube | |
Test Tube Rack with Holder |