missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | HOOC(CH2)4COOH |
Beilstein | 1209788 |
Solubility Information | Freely soluble in methanol, boiling water, cyclohexane, acetone, water, ethanol. Insoluble in benzene,petroleum ether. |
Formula Weight | 146.14g/mol |
Percent Purity | 99.6 to 101% (calc. to the dried substance) |
Quantity | 100 g |
Flash Point | 196°C (384.8°F) |
Ignition Point | 788°F |
Residue after Ignition | ≤0.002% (as SO4) |
Vapor Density | 5 (vs air) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'