Learn More
3M™ Adhesive Transfer Tape
3M™ Adhesive Transfer Tape is ideal for bonding textured LSE plastics, powder coatings, and oily metals.
Brand:  3M™ 7100089548
Product Code. 15763518
  • 13615.00 NOK / 1 roll
Only null left
Estimated Shipment: 07-08-2025
Description
  • Bond textured LSE plastics, powder coatings and oily metals
  • Apply good chemical and moisture resistance
  • Maintain moisture stability
  • Good resistance to industrial chemicals, consumer chemicals, moisture and humidity
  • Ideal solution for general purpose attachment and assemblies of a wide variety of materials
  • Meets a wide variety of automotive and OEM specifications
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ Adhesive Transfer Tape > Length: 55 m