missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5122271
This item is not returnable.
View return policy
Description
This gene encodes a member of the membrane-bound adenylyl cyclase enzymes. Adenylyl cyclases mediate G protein-coupled receptor signaling through the synthesis of the second messenger cAMP. Activity of the encoded protein is stimulated by the Gs alpha subunit of G protein-coupled receptors and is inhibited by protein kinase A, calcium and Gi alpha subunits. Single nucleotide polymorphisms in this gene may be associated with low birth weight and type 2 diabetes. Alternatively spliced transcript variants that encode different isoforms have been observed for this gene. [provided by RefSeq, Dec 2010].Specifications
Adenylate Cyclase 5 | |
Polyclonal | |
Unconjugated | |
ADCY5 | |
AC5; Adcy5; adenylate cyclase 5; adenylate cyclase type 5; Adenylate cyclase type V; adenylyl cyclase 5; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; AW121902; FDFM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
111 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O95622 | |
ADCY5 | |
Gly167-Glu385 (Accession O95622), with N-terminal His Tag | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |