missing translation for 'onlineSavingsMsg'
Learn More
Adenosine-5'-Diphosphate Trilithium Salt Hydrate, MP Biomedicals™ Product Code.: 11433081

Adenosine-5'-Diphosphate Trilithium Salt Hydrate, MP Biomedicals™

Product Code. 11433081 Shop All MP Biomedicals Products
500 mg
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Product Code. 11433081

Brand: MP Biomedicals 02150261.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adenosine-5'-Diphosphate Trilithium Salt Hydrate, MP Biomedicals is a lithium salt formulation of the essential metabolic compound ADP.

Adenosine 5′-diphosphate has been used to study the release, neuronal effects and removal of extracellular β-nicotinamide adenine dinucleotide (β-NAD+) in the rat brain. Adenosine 5′-diphosphate has also been used to compare prasugrel versus clopidogrel antiplatelet therapy after acute coronary syndrome.

TRUSTED_SUSTAINABILITY

Specifications

CAS 31008-64-7
Molecular Formula C10H12Li3N5O10P2
Refractive Index ∼1.90 (Predicted) (Lit.)
MDL Number MFCD00065469
Quantity 500 mg
Synonym adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
InChI Key LZGPPAHUZSOGHJ-DJXXCMMGNA-K
SMILES [Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
IUPAC Name trilithium(1+) ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Weight (g/mol) 445.00
PubChem CID 56841973
Formula Weight 445 (anhydrous)
Percent Purity ≥96%
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adenosine-5'-Diphosphate Trilithium Salt Hydrate, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.