missing translation for 'onlineSavingsMsg'
Learn More
Adenosine-5'-diphosphate trilithium salt, 96 to 99%, For HPLC analysis, MP Biomedicals™ Product Code.: 11453081

Adenosine-5'-diphosphate trilithium salt, 96 to 99%, For HPLC analysis, MP Biomedicals™

Product Code. 11453081 Shop All MP Biomedicals Products
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11453081

Brand: MP Biomedicals 0215026105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 31008-64-7
Molecular Formula C10H12Li3N5O10P2
Molecular Weight (g/mol) 445.00
MDL Number MFCD00065469
InChI Key LZGPPAHUZSOGHJ-DJXXCMMGNA-K
Synonym adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
PubChem CID 56841973
IUPAC Name trilithium(1+) ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 96 to 99%
Quantity 5 g
Formula Weight 454
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphosphate trilithium salt, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adenosine-5'-diphosphate trilithium salt, 96 to 99%, For HPLC analysis, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.