Learn More
Memmert™ Additional Stainless Steel Grid for UF750, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ UF750B32190
6715.10 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
UF750 Oven | |
Reinforced Grid | |
Stainless Steel | |
60 kg |
Product Content Correction
Product Title
Memmert™ Additional Stainless Steel Grid for UF750, Reinforced Chamber Option > For Use With: UF750