Learn More
Memmert™ Additional Stainless Steel Grid for SN160plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ SN160plusE29767
This item is not returnable.
View return policy
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
SN160plus Sterilizer | |
Reinforced Grid | |
Stainless Steel |
Product Content Correction
Product Title
Memmert™ Additional Stainless Steel Grid for SN160plus, Reinforced Chamber Option > For Use With: SN160plus