missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Adapters with NS Stopcock, Bent
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05105003
This item is not returnable.
View return policy
Description
- Adapters with NS socket
- Bent glass olive (90°) and NS stopcock with solid glass plug
- Bore: 2,5 mm
- Made of DURAN™ tubing
Specifications
Bent glass olive (90°) | |
Borosilicate Glass (3.3) | |
NS 24/29 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasintrumente Adapters with NS Stopcock, Bent > NS 24/29
Spot an opportunity for improvement?Share a Content Correction