Learn More
SI Analytics™ Conversion Adapter
SI Analytics™ Conversion Adapter transforms all pH meters with DIN sockets into BNC plugs.
Brand:  SI Analytics™ 285205006
Product Code. 10334911
  • 419.00 NOK / Each
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Adapter for DIN socket
  • Converts socket in BNC plug
  • For conversion of all pH-meters with DIN entrance into BNC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conversion Adapter
Adapter DIN socket or BNC Connector
Conversion of all pH-meters with DIN entrance to BNC