Learn More
Eppendorf™ Adapter for HPLC Vials
Designed for use with Eppendorf Rotor FA-45-20-17
Brand: Eppendorf™ 5820770007
476.90 NOK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adapter for 1 HPLC Vial
- For Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
- Available in packs of 4
Specifications
Adapter | |
Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17 | |
HPLC | |
1/Pk. |
Product Content Correction
Product Title
Eppendorf™ Adapter for HPLC Vials > For Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17