Learn More
Hettich™ Adapter for Carrier no. 1427
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5273
 View more versions of this product
Product Code. 10021942
  • 1180.00 NOK / Each
Description
  • Adapter for Carrier no. 1427
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
9 x 1.5 to 2 mL Tubes
38 mm
9
Microtube
11 mm
Carrier no. 1427
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Adapter for Carrier no. 1427 > 6 x 15mL