Learn More
Eppendorf™ Adapter
Brand:  Eppendorf™ 4960832006
760.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10378271
  • 763.04 NOK / Each
Only null left
In Stock
Specifications
Adapter
1 Ea.
45 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Adapter > Thread Size: 45mm