Learn More
Tocris Bioscience™ ACT 462206
Potent, dual orexin receptor antagonist
Brand: Tocris Bioscience™ 5319/50
5812.38 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
ACT 462206 is a potent, dual orexin receptor antagonist (IC50 values are 11 and 60 nM for OX2 and OX1 receptors respectively). Exhibits anxiolytic-like properties in vivo. Brain penetrant.Specifications
>99% | |
ACT 462206 | |
ACT 462206 |
50 mg | |
388.48 |
For Research Use Only