missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17138-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
ACSS1 | |
Polyclonal | |
Unconjugated | |
ACSS1 | |
ACAS2L, AceCS2, AceCS2L, Acetate CoA ligase 2, Acetyl CoA synthetase 2, ACSS1, KIAA1846 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296259, 68738, 84532 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99NB1, Q9NUB1 | |
Acss1 | |
ACSS1 Fusion Protein Ag10896 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |