missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16140-1-AP-150UL
This item is not returnable.
View return policy
Description
Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA. Has some preference toward medium-chain substrates. Plays a role in adipocyte differentiation.Specifications
ACSF2 | |
Polyclonal | |
Unconjugated | |
ACSF2 | |
ACSF2, ACSMW, AVYV493 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
264895, 619561, 80221 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q499N5, Q8VCW8, Q96CM8 | |
Acsf2 | |
ACSF2 Fusion Protein Ag9199 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |