Learn More
Fisherbrand™ Acrylic Racks for Autosampler Vials
Easy handling and transportation of sample vials
859.00 NOK - 1020.00 NOK
Specifications
Product Type | Vial Rack |
---|---|
Closure Included | No |
Autoclavable | Not Autoclavable |
Color | Transparent |
Material | Acrylic |
Product Code | Brand | Dimensions (L x W x H) | Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Positions | Price | Quantity & Availability | |||||
11767597
|
Fisherbrand™
12 21 1001 |
173 x 95 x 20mm | 50 |
890.00 NOK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11728256
|
Fisherbrand™
15 21 1664 |
175.8 x 115.5 x 20mm | 40 |
925.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11708256
|
Fisherbrand™
08 21 1000 |
173 x 91 x 20mm | 50 |
859.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
11738256
|
Fisherbrand™
24 21 1002 |
160 x 160 x 30mm | 25 |
1020.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Vial-Rack
- Acrylic
- Transparent acrylic material makes filling visible
- Stable standing position
Specifications
Vial Rack | |
Not Autoclavable | |
Acrylic |
No | |
Transparent |
Product Title
Fisherbrand™ Acrylic Racks for Autosampler Vials