Learn More
Fisherbrand™ Acrylic Racks for Autosampler Vials
859.75 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Vial-Rack
- Acrylic
- Transparent acrylic material makes filling visible
- Stable standing position
Specifications
Vial Rack | |
Not Autoclavable | |
Acrylic | |
4mL shell vials | |
175.8 x 115.5 x 20mm |
No | |
Transparent | |
40 | |
1 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%