missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
- Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
- It polymerizes in the presence of free radicals in aqueous solutions.
- It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
- Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.

Specifications
Specifications
Physical Form | Crystalline Powder |
Density | 1.127g/cm3 at 25°C |
Color | White |
Quantity | 500 g |
Assay Percent Range | ≥99% |
Formula Weight | 71.1 |
Chemical Name or Material | Acrylamide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acrylamide, ≥99%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction