Learn More
Thermo Scientific Chemicals Glass beads, 400-800 micron
Brand:  Thermo Scientific Chemicals 397640250
350.96 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10264954
  • 361.00 NOK / 25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass
25 g
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471741 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number397640250,397641000,397645000
Lot NumberA0472021 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number397640250,397641000,397645000
Lot NumberA0463208 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number397640250,397641000,397645000
Lot NumberA0458519 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number397640250,397641000,397645000
Lot NumberA0450372 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number397640250,397641000,397645000
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Chemicals Glass beads, 400-800 micron > 25g; Glass bottle