Learn More
Cytiva AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
Brand:  Cytiva 12991
1637.39 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 17104251
  • 2040.00 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Edge
Description
Description
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrophilic PES
Sterile
1/Ea.
0.8/0.2 μm
500 cm2
Stepped Hose Barb
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Cytiva AcroPak™ 500 Capsules with Supor™ Membrane, Sterile > 0.8 μm/0.2 μm