missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Aconitase 2 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16808013
|
Proteintech
11134-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16898003
|
Proteintech
11134-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.Specifications
Aconitase 2 | |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q99798, Q99KI0, Q9ER34 | |
11429, 50, 79250 | |
Aconitase 2 Fusion Protein Ag1590 | |
Primary | |
-20°C | |
ACO2 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACO2, Aconitase, aconitase 2, aconitase 2, mitochondrial, ACONM, Citrate hydro lyase | |
ACO2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |