missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, Supragradient HPLC Grade, Scharlab Product Code.: 11389783

Acetonitrile, Supragradient HPLC Grade, Scharlab

Product Code. 11389783
2.5 L, Amber Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11389783

Brand: Scharlab AC03312500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guaranteed specifications include excellent high UV transmission and minimum peak impurities in gradient HPLC. Acetonitrile, Supragradient HPLC Grade, Scharlab records fluorescence at EX wavelengths of 220-450nm and EM wavelength of 250-550nm.

This HPLC-grade solvent is of appropriate purity to be used as a mobile phase in HPLC in analytical and preparative separations.

  • Undergoes highly efficient distillation, solid phase absorption and other treatments to eliminate impurities that could interfere in HPLC analysis
  • Microfiltered and packed in amber-coloured bottles (sealed with a PTFE membrane in the cap) to avoid possible contamination

Specifications

Melting Point -45.7°C
Absorbance 0.097AU at 195nm, 0.022AU at 200nm, 0.013AU at 210nm, 0.009AU at 220nm
CAS 75-05-8
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0001meq/g
Density 0.78g/cm3
Boiling Point 81.6°C
Quantity 2.5 L
Flash Point 2°C
Identification Passes test
Ignition Point 524°C
Packaging Amber Glass Bottle
Physical Form Liquid
Refractive Index 1.3442
Residue after Evaporation Max. 0.0001%
Molecular Formula C2H3N
Linear Formula CH3CN
Vapor Pressure 97 hPa at 20°C
Water Max. 0.01%
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Solubility Information Miscible in water
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05g/mol
Percent Purity Min. 99.9%
Grade Supragradient HPLC
Chemical Name or Material Acetonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, Supragradient HPLC Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.