missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, LC-MS, Scharlab
Click to view available options
Quantity:
1 L
Specifications
Specifications
CAS | 75-05-8 |
Quantity | 1 L |
Molecular Formula | C2H3N |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Molecular Weight (g/mol) | 41.05 |
Grade | LC-MS |
Safety and Handling
Product Identifier
- Acetonitrile, LC-MS
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'