missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, Gradient (240nm), HPLC Grade, Scharlab Product Code.: 11319803
compliance-icons compliance-icons

Acetonitrile, Gradient (240nm), HPLC Grade, Scharlab

Product Code. 11319803
2.5 L, 2.50 L
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
This item is not returnable. View return policy

Product Code. 11319803

Brand: Scharlab AC03292500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used primarily in chromatography, synthesis of organic products, and in solvents.

Microfiltered through membranes of pore diameter 0.22ÎĽm and gradient, far-UV, HPLC grade.

  • Synonyms: Methyl cyanide, Cyanomethane
  • Molecular Formula: CH3CN
  • Molar Mass: 41.05g/mol
  • CAS: 75-05-8
  • EIN ECS No.: 200-835-2
  • Dielectric Constant: 37.5 at 20°C
  • Reducing Substances: Passes test

Specifications

Melting Point -45.7°C
Absorbance Max background: 0.01AU at 240nm
CAS 75-05-8
Color Colorless
Acidity 0.0002mEq/g max
Alkalinity 0.0001mEq/g max
Density 0.786g/cm3
Boiling Point 81.6°C
Quantity 2.5 L
Flash Point 2°C
Identification Passes Test (IR spectrum)
Ignition Point 524°C
Physical Form Liquid
Refractive Index 1.3442 (n 20°C/D)
Residue after Evaporation 0.0002% max
Molecular Formula C2H3N
Vapor Pressure 97hPa at 20°C
Water 0.02% max
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Solubility Information Miscible in water (20°C)
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05g/mol
Percent Purity 99.9% min
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile, gradient 240nm/ far UV HPCL grade
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, Gradient (240nm), HPLC Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.