missing translation for 'onlineSavingsMsg'
Learn More
Acetone, VLSI Grade, Scharlab Product Code.: 11349783

Acetone, VLSI Grade, Scharlab

Product Code. 11349783
25 L, Stainless Steel Drum
Click to view available options
Quantity:
25 L
Packaging:
Stainless Steel Drum
This item is not returnable. View return policy

Product Code. 11349783

Brand: Scharlab AC0320025B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetone, VLSI Grade, Scharlab is guaranteed to be free of water, metals and particles.

VLSI (Very Large Scale Integration) Grade solvents are often used in the semiconductor industry and in electronic industry production, mainly in the cleaning of integrated circuits.

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Acidity Max. 0.0003meq/g
Alkalinity Max. 0.0006meq/g
Density 0.79g/cm3 at 20°C
Boiling Point 56°C
Quantity 25 L
Flash Point −20°
Identification Passes test
Ignition Point 540°C
Odor Pungent
Packaging Stainless Steel Drum
Physical Form Liquid
Refractive Index 1.3588
Residue after Evaporation Max. 0.0005%
Molecular Formula C3H6O
Linear Formula (CH3)2CO
MDL Number MFCD00008765
Vapor Pressure 233 hPa at 20°C
Water Max. 0.1%
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Miscible in water
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity Min. 99.5%
Grade LCSI
Chemical Name or Material Acetone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, VLSI Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.