missing translation for 'onlineSavingsMsg'
Learn More
Acetone, Pharmpur™, Scharlab Product Code.: 11339773
compliance-icons compliance-icons

Acetone, Pharmpur™, Scharlab

Product Code. 11339773
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 11339773

Brand: Scharlab AC0312005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in solvents, analytical chemistry, synthesis of organic products, and photography.

  • Synonyms: Dimethyl ketone, 2-Propanone
  • Molecular Formula: C3H6O
  • Molar Mass: 58.08g/mol
  • CAS: 67-64-1
  • EIN ECS No.: 200-662-2
  • Dielectric Constant: 20.7 at 25°C
  • Reducing Substances: Passes test

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Appearance Passes Test
Density 0.79g/cm3
Boiling Point 56°C
Quantity 5 L
Flash Point < −20°C
Identification Passes Test
Ignition Point 540°C
Physical Form Liquid
Refractive Index 1.3488 (n 20°C/D)
Residue after Evaporation 0.002% max
Molecular Formula C3H6O
MDL Number MFCD00008765
Vapor Pressure 233hPa at 20°C
Water 0.3% max
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Miscible in water (20°C)
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity 99.5% min
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone, extra pure, Pharmpur, Ph Eur, BP, NF
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, Pharmpur™, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.