missing translation for 'onlineSavingsMsg'
Learn More
Acetone, Multisolvent™ Grade, Sharlab Product Code.: 11339783

Acetone, Multisolvent™ Grade, Sharlab

Product Code. 11339783
2.5 L, Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 11339783

Brand: Scharlab AC03102500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used for almost every laboratory application. Acetone, Multisolvent™ Grade from Sharlab features high purity and low water content for use in applications requiring an HPLC-grade, ACS-grade, ISO-grade, UV-VIS-grade or Reagent-grade product.

Multisolvent™ products are most frequently use for HPLC, but can also be used for numerous other applications, making it one of the most universal grades available. These versatile products have numerous advantages over application-specific solvents:

  • Guarantee results: QC controls are performed by modern analytical techniques for all guaranteed applications
  • Optimize purchasing and stocking: One product for most applications prevents the need to order, stock and track the supply of multiple grade
  • Optimal packaging: Highly-resistant and durable amber glass bottles or stainless steel drums ensure the highest product safety and quality
  • Lower costs by unifying: Multiple-use products at single-grade prices to avoid buying small quantities of specific products
  • Preserve the environment: Returnable packaging (one-way glass bottles or SS returnable drums) help prevent waste

Specifications

Melting Point -95°C
Absorbance 1.000 at 330nm, 0.301 at 335nm, 0.097 at 339nm, 0.046 at 342nm, 0.009 at 350nm
CAS 67-64-1
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0002meq/g
Appearance Passes test
Density 0.79g/cm3 at 20°C
Boiling Point 56°C
Quantity 2.5 L
Flash Point −20°
Identification Passes test
Ignition Point 540°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3588
Residue after Evaporation Max. 0.0002%
Molecular Formula C3H6O
Linear Formula (CH3)2CO
MDL Number MFCD00008765
Vapor Pressure 233 hPa at 20°C
UN Number 1090
Water Max. 0.2%
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity Min. 99.8%
Grade Applications: solvents, analytical chemistry, synthesis of organic products, photography
Chemical Name or Material Acetone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, Multisolvent™ Grade, Sharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.