missing translation for 'onlineSavingsMsg'
Learn More

Acetone, MP Biomedicals™

Acetone ACS, ≥99.5%, Clear, Colorless Liquid

377.00 NOK - 1495.00 NOK

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11442451
View Documents
MP Biomedicals
0219383280
100 mL
436.00 NOK
100mL
Please to purchase this item. Need a web account? Register with us today!
11456221
View Documents
MP Biomedicals
0219383290
0.5 L
377.00 NOK
500mL
Please to purchase this item. Need a web account? Register with us today!
11466221
View Documents
MP Biomedicals
0219383291
1 L
450.00 NOK
1 L
Please to purchase this item. Need a web account? Register with us today!
11438262
View Documents
MP Biomedicals
0219383204
4 L
1495.00 NOK
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • It is a commonly used material in the manufacture of various chemicals like methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid (via the ketene process), iodoform, and bromoform.
  • Other products where acetone is utilized in manufacture include rayon, photographic films, and paint and varnish removers.
  • In chemical synthesis, acetone has been utilized in the synthesis and isolation of both organic and inorganic compounds and complexes.
  • These include t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles, and trispyrazolylborate platinum compounds.
  • Acetone is also used in HPLC analysis of organic and inorganic species, including gold nanoparticle clusters and plant oils. Acetone has been utilized in protein crystallization.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : AL3150000

missing translation for 'storageNote' : Store at Room Temperature(15- 30°C).

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, MP Biomedicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.