missing translation for 'onlineSavingsMsg'
Learn More
Acetone, for GC Residue Analysis, Scharlab Product Code.: 11319783

Acetone, for GC Residue Analysis, Scharlab

Product Code. 11319783
2.5 L, Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 11319783

Brand: Scharlab AC03082500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the preparation of specimens in the analysis of pesticides where solvent extraction is required. Acetone, for GC Residue Analysis, Scharlab is an extraordinarily pure solvent, with controls that ensure the absence of peaks of over 3 ppt.

GC Residue Analysis Grade solvents are used to prepare specimens for pesticide analysis. The sample is evaporated, the residue is re-dissolved in the solvent and the solution is analyzed using gas chromatography. Evaporation concentrates both the residue and any impurities in the solvent itself, so this type of analysis requires extraordinarily pure solvents.

  • Controls performed by GC-ECD to ensure the absence of peaks of over 3 ppt (expressed as lindane), from low, medium and high boiling points (from 1,2,4-trichlorobenzene to decachlorobiphenyl), and also in the area around that of 2,4,5-trichlorobiphenyl.
  • These controls ensure that the quality of GC Residue Analysis solvents is suitable for organochlorinated, dioxin, furan and PCB pesticide residue analysis.
  • In pesticide residue analaysis, anhydrous sodium sulfate is normally used as a drying agent once the extraction process is performed, after separating the aqueous and organic phases.

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 56°C
Quantity 2.5 L
Flash Point −20°
Identification Passes test
Ignition Point 540°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3588
Residue after Evaporation Max. 0.0001%
Molecular Formula C3H6O
Linear Formula (CH3)2CO
MDL Number MFCD00008765
Vapor Pressure 233 hPa at 20°C
UN Number 1090
Water Max. 0.2%
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Miscible in water
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity Min. 99.8%
Grade Suitable for organohalogenated pesticide and dioxins, furans and PCBs residue analysis
Chemical Name or Material Acetone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, for GC Residue Analysis, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.