Learn More
Acetone-d6, for NMR, 99+ atom % D
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 0.8700g/mL |
Deuterium (D) | 99.0% min. |
Flash Point | −20°C |
Packaging | Glass Bottle |
Quantity | 10 mL |
Melting Point | -93.0°C |
Boiling Point | 55.0°C |
Linear Formula | CD3COCD3 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0469953 | Certificate of Analysis | 17/05/2025 | 174900500,174900100 |
A0472299 | Certificate of Analysis | 17/05/2025 | 174900500,174900100 |
A0470276 | Certificate of Analysis | 17/05/2025 | 174900500,174900100 |
A0472130 | Certificate of Analysis | 17/05/2025 | 174900500,174900100 |
A0472148 | Certificate of Analysis | 17/05/2025 | 174900500,174900100 |
Safety and Handling


- Acetone-d6
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.