Learn More
Lancer™ LancerAcid LCA-A Rinse
Lancer's most commonly used rinsing agent, LCA-A is an acetic acid-based neutralizer solution for excellent rinsing on all standard laboratory soils, organics, pharmaceutical-based contaminants, petroleum and other difficult soils.
Brand: Lancer™ LCA25
This item is not returnable.
View return policy
Description
- Made specifically for use in all high pressure washers
- Premixed for direct use from the bottle to the washer
- Dosing is automatic by peristaltic pump
- Manufactured with dedicated production lines and a double filtration system to ensure integrity
- Phosphate-free and non-foaming
Specifications
Acetic Acid-based Neutralizer | |
Provides an excellent rinse on all standard laboratory soils and contaminants | |
2.5 gal. |
Double filtered | |
High pressure glassware washers |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P264-Wash thoroughly after handling.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Acetic Acid