Learn More
Lancer™ LancerAcid LCA-A Rinse
Lancer's most commonly used rinsing agent, LCA-A is an acetic acid-based neutralizer solution for excellent rinsing on all standard laboratory soils, organics, pharmaceutical-based contaminants, petroleum and other difficult soils.
Brand:  Lancer™ LCA25
Product Code. 13142230
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made specifically for use in all high pressure washers
  • Premixed for direct use from the bottle to the washer
  • Dosing is automatic by peristaltic pump
  • Manufactured with dedicated production lines and a double filtration system to ensure integrity
  • Phosphate-free and non-foaming
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acetic Acid-based Neutralizer
Provides an excellent rinse on all standard laboratory soils and contaminants
2.5 gal.
Double filtered
High pressure glassware washers
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash thoroughly after handling.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Acetic Acid