Learn More
Acetic Acid Solution, 0.1mol/L, 1000mL, Fisher Chemical™ Product Code.: 12970904
Acetic Acid Solution, 0.1mol/L, 1000mL, Fisher Chemical™
BIOPHARMA
Product Code. 12970904 Shop All Fisher Chemical Products
1000 mL
Click to view available options
Quantity:
1000 mL
Product Code. 12970904
Brand: Fisher Chemical VL 200 1000
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System; also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 0.1mol/L
Quantity 1000 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1097754 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberVL 200 1000
Lot Number1032166 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 200 1000
Lot Number1019197 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 200 1000
Product Content Correction
Product Title
Acetic Acid Solution, 0.1mol/L, 1000mL, Fisher Chemical™ >