missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 10% vol. Solution, Chem-Lab
Click to view available options
Quantity:
1 L
Packaging:
PE
Specifications
Specifications
CAS | 64-19-7 |
Molecular Formula | C2H4O2 |
MDL Number | MFCD00036152 |
UN Number | UN2790 |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
Concentration | 100mL CH3COOH / 1L H2O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
Show More |
Safety and Handling


Product Identifier
- Acetic acid 10 vol. % solution
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Acetic acid glacial
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetic Acid, 10% vol. Solution, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction