missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ACD |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16804415
|
Proteintech
25849-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16894405
|
Proteintech
25849-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a uridine 5′ -monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5′ -monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5′ -monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.Specifications
ACD | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96AP0 | |
65057 | |
ACD Fusion Protein Ag23021 | |
Primary | |
-20°C | |
ACD |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACD, PIP1, PTOP, TINT1, TPP1 | |
ACD | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |