Learn More
Fisherbrand™ accuSpin 21R Refrigerated Microcentrifuge
GREENER_CHOICE
The Fisherbrand accuSpin 21 Microcentrifuges with a 24 x 1.5/2.0 mL rotor and a ClickSeal biocontainment lid that offers exceptional acceleration of up to 21,000 x g in as little as 12 seconds.
Brand:  Fisherbrand™ 75003249
50044.98 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 17616315
  • 59962.10 NOK / 1 set
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
  • Durable and flexible rotors
  • Outstanding corrosion resistance with lightweight engineered polymer rotors
  • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns
  • ClickSeal™ Biocontainment solution
  • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids
  • Quiet, Easy to Use Design
  • Simplified operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
  • Sleek, lightweight, space-saving design
  • Durable and easy to clean and maintain
  • Quiet performance promotes a safe, stress-free environment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
21,000 x g
440 mm
24 x 1.5/2 mL rotor with ClickSeal Biocontainment Lid
Yes
14,800 rpm
330 mm
292 mm
230 V
Yes
Videos
  • Fisherbrand - POTM - August En
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand - POTM - August En
    SDS
    Documents Certificates
    Promotions
    Promotions
    • Thermo Scientific Sorvall , Fisherbrand , Thermo Scientific
      Unlock special pricing on top centrifuges. Make us an offer today and upgrade your lab equipment. Limited time only!
      Promo Code: 21629
      Expires2025-12-31
      Days
      187
      Hours
      8
      Minutes
      56
      Seconds
      23
    • Multiple Manufacturers
      Get amazing savings on products for your analyses
      Promo Code: 24135
      Expires2025-07-13
      Days
      17
      Hours
      8
      Minutes
      56
      Seconds
      23
    • Multiple Manufacturers
      Save up to 59% on some of our most popular products
      Promo Code: 24140
      Expires2025-08-24
      Days
      58
      Hours
      8
      Minutes
      56
      Seconds
      23
    • Multiple Manufacturers
      Save big on essential products for cancer research
      Promo Code24147
      Expires2025-07-31
    • Multiple Manufacturers
      Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
      Promo Code: 24156
      Expires2025-06-30
      Days
      3
      Hours
      8
      Minutes
      56
      Seconds
      23
      Hot Deal
      *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
      Terms & Conditions See Full Promo Details
    • Multiple Manufacturers
      Browse exclusive prices for our academic customers
      Promo Code: 25081
      Expires2025-12-31
    Product Content Correction
    Product Title
    Fisherbrand™ accuSpin 21R Refrigerated Microcentrifuge > Refrigerated