Learn More
Invitrogen™ AccuPrime™ SuperMix I
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.
Brand:  Invitrogen™ 12342028
 View more versions of this product
Product Code. 10297522
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix I sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.
In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.
AccuPrime SuperMix I is designed for amplification of genomic DNA amplicons (> 200 bp), plasmid DNA, or cDNA templates. Alternatively, AccuPrime SuperMix II is designed to amplify a broader range of amplicon sizes up to 4 kb.
• Built-in hot start
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AccuPrime SuperMix I is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 1000 amplification reactions of 25 μL each are provided. AccuPrime SuperMix I Components: 40 mM Tris-HCl (pH 8.4), 100 mM KCl, 3 mM MgCl2, 400 μM dGTP, 400 μM dATP, 400 μM dTTP, 400 μM dCTP, AccuPrime Taq DNA Polymerase, thermostable AccuPrime protein, stabilizers.
1000 Reactions: 1 x 12.5 mL
Store at -20°C in a non-frost-free freezer.
Low
2X
1000 Reactions