Learn More
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes with these accessories.
Brand:  Thermo Scientific™ 910006-WA
 View more versions of this product
Product Code. 11737767
  • 1963.47 NOK / Pack of 3
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
Estimated Shipment: 29-07-2025
Description
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
  • Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
  • Designed for glass body pH electrodes
  • Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Electrode storage bottles
6 mm Electrode, Glass
Electrode Storage Bottle
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Orion™ pH Electrodes >