Learn More
Fisherbrand™ Accessories for Fisherbrand™ RT Basic Series Magnetic Stirrers
Optimize performance and flexibility
219.00 NOK - 233.00 NOK
Specifications
For Use With (Equipment) | RT Basic Series Magnetic Stirrer |
---|---|
Type | White Silicone Plate Cover |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
15316617
|
Fisherbrand™
88860145 |
170 mm |
219.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
15326617
|
Fisherbrand™
88860146 |
220 mm |
233.00 NOK
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
Description
- White
- Silicone
Specifications
RT Basic Series Magnetic Stirrer | |
White Silicone Plate Cover |
Product Title
Fisherbrand™ Accessories for Fisherbrand™ RT Basic Series Magnetic Stirrers