Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker Product Code.: 15564110
Estimated Shipment: 10-04-2026
  • ONLINE EXCLUSIVE PRICE
    489.00 NOK
    460.93 NOK / Each
    Save 28.07 NOK
    5% Off
Only null left
Add to basket
Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker
Product Code. 15564110 Shop All Fisherbrand Products
Infusion Bottle Clamp, 500 mL Infusion Bottle, Each
Click to view available options
Product Type:
Adjustable Tube Rack
Fixed Tube Rack
Flask Clamp
Funnel Clamp
Infusion Bottle Clamp
Platform with Adjustable Bars
Platform with Clamps
Platform with Rubber Mat
Holds:
1000 mL Flask
1000 mL Infustion Bottle
12 x 100 mL Flasks, 150 mL Beakers
2000 mL Flask
3000 mL Flask
40 x 14 mm dia. Tubes
40 x 16 mm dia. Tubes
500 mL Flask
500 mL Infusion Bottle
5000 mL
6 x 250 mL Flasks, 150 mL Beakers
Funnels
Vessels
Unit Size:
Each
Product Code. 15564110
  • ONLINE EXCLUSIVE PRICE
    489.00 NOK
    460.93 NOK / Each
    Save 28.07 NOK
    5% Off
Only null left
Estimated Shipment: 10-04-2026
Add to basket
Only null left
Add to basket
Accessories required for the use of Fisher Scientific™ Multi Platform Shaker
Accessories for multi platform shaker to enhance performance and flexibility
  • Increase performance and flexibility
TRUSTED_SUSTAINABILITY
Specifications
Holds 500 mL Infusion Bottle
Product Type Infusion Bottle Clamp
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker >