Learn More
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Optimize performance and flexibility
Brand:  Fisherbrand™ 88860148
97.68 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15346617
  • 116.00 NOK / Each
In Stock
to see stock.
Explore more special offers
Description
Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers. Optimize performance and flexibility
  • Accessory for use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
C-5, Clamp Holder (PP body, 12mm dia.)
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    44
    Hours
    1
    Minutes
    36
    Seconds
    12
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers > C-5, Clamp Holder (PP body, 12mm dia.), Available for RT2 Advanced Hotplate Stirrer only