Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
Brand: Thermo Scientific™ CIC0001830
273.73 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
- Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for stirring
- Raised display design protects electronics from spills
- Optional splash guard protection shield
- Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
LED | |
Supporting Rod |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%