Learn More
Thermo Scientific™ Accessories for Bottle/Tube Roller
Use these accessories to optimize the performance of your Thermo Scientific™ Bottle/Tube Roller.
Brand: Thermo Scientific™ 88881012
47.49 NOK valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Description
Use Thermo Scientific™ Accessories for Bottle/Tube Roller to optimize flexibility and performance.
Specifications
15 mL | |
13.2 x 2.65 mm (0.52 x 0.1 in.) | |
Bottle/Tube Roller |
O-ring for 15 mL tube | |
O-ring for 15mL tube (pack of 20) 13.2 x 2.65mm 0.52 x 0.1 in. |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.