Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Access Port with silicone plug is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Brand: BINDER™ 8012-2077
11312.72 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Access Port
- With Silicone Plug
- For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Access port with silicone plug - 100 mm, left | |
Steel | |
Left Position | |
3.9 in. |
BINDER Chambers Series KBF-S and KBF-S ECO | |
Access Port | |
Silicone plug | |
100 mm |
Product Content Correction
Product Title
BINDER™ Access Port with Silicone Plug > Diameter: 100 mm, Position: left