Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Access Port with silicone plug is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Brand:  BINDER™ 8012-2075
11312.72 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17373663
  • 12445.00 NOK / Each
Only null left
Estimated Shipment: 10-09-2025
Description
  • Access Port
  • With Silicone Plug
  • For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Specifications
Access port with silicone plug - 100 mm, top
Steel
Top Position
3.9 in.
BINDER Chambers Series KBF-S and KBF-S ECO
Access Port
Silicone plug
100 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Access Port with Silicone Plug > Diameter: 100 mm, Position: top