Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Access Port with silicone plug is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Brand:  BINDER™ 8012-2074
 View more versions of this product
Product Code. 17363663
  • 7775.00 NOK / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Description
  • Access Port
  • With Silicone Plug
  • For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Specifications
Access port with silicone plug - 50 mm, top
Steel
Top Position
2 in.
BINDER Chambers Series KBF-S and KBF-S ECO
Access Port
Silicone plug
50 mm
Product Title
BINDER™ Access Port with Silicone Plug > Diameter: 50 mm, Position: top