Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Access Port with silicone plug is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Brand: BINDER™ 8012-2073
6059.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Access Port
- With Silicone Plug
- For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
Access port with silicone plug - 30 mm, top | |
Steel | |
Top Position | |
1.2 in. |
BINDER Chambers Series KBF-S and KBF-S ECO | |
Access Port | |
Silicone plug | |
30 mm |
Product Content Correction
Product Title
BINDER™ Access Port with Silicone Plug > Diameter: 30 mm, Position: top