missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20941-1-AP-20UL
This item is not returnable.
View return policy
Description
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.Specifications
ACBD4 | |
Polyclonal | |
Unconjugated | |
ACBD4 | |
ACBD4, HMFT0700 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79777 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NC06 | |
ACBD4 | |
ACBD4 Fusion Protein Ag15045 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction