Learn More
AC710, MedChemExpress
MedChemExpress AC710 is a potent PDGFR inhibitor with Kds of 0.6, 1.57, 1, 1.3, 1.0 nM for FLT3, CSF1R, KIT, PDGFRα and PDGFRβ, respectively.

2117.70 NOK - 18284.93 NOK
Specifications
Chemical Name or Material | AC710 |
---|---|
CAS | 1351522-04-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C31H42N6O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496196
![]() |
Medchem Express
HY-13493 |
100 mg |
18284.93 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16485708
![]() |
Medchem Express
HY-13493 |
10 mg |
3429.41 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16418407
![]() |
Medchem Express
HY-13493 |
5 mg |
2117.70 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401877
![]() |
Medchem Express
HY-13493 |
50 mg |
11426.10 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AC710 is a potent PDGFR inhibitor with Kds of 0.6, 1.57, 1, 1.3, 1.0 nM for FLT3, CSF1R, KIT, PDGFRα and PDGFRβ, respectively.Specifications
AC710 | |
White | |
C31H42N6O4 | |
O=C(C1=NC=C(OC2CC(C)(C)N(CC)C(C)(C)C2)C=C1)NC3=CC=C(NC(NC4=NOC(C(C)(C)C)=C4)=O)C=C3 | |
562.7 | |
Research | |
Cancer-Kinase/protease |
1351522-04-7 | |
Research | |
DMSO : 14 mg/mL (24.88 mM; Need ultrasonic and warming) | |
562.7 | |
99.89% | |
Solid |
Safety and Handling
H315∣H317∣H318∣H334∣H335∣H341∣H361∣H370∣H413
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.