Learn More
Fisherbrand™ MagLab™ MagBox™ Storage Box
Create storage space without crowding limited workspace. Fisherbrand™ MagLab™ MagBox™ Storage Box uses magnetic mounting to store miscellaneous lab and office supplies off the benchtop. Constructed of durable, brightly colored ABS plastic.
Brand: Fisherbrand™ HS23504
197.54 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
Create lab storage space off your bench top with the MagLab™ storage system.- Easily organize common lab and office accessories off your bench top
- Robust ABS plastic is translucent for easy content identification and access
- Features four powerful magnets for a secure hold onto most metal surfaces
- Mount on non-metallic surfaces with a self-adhesive metal mounting plate, provided
Specifications
Storage Container | |
ABS | |
130 x 78 x 102 mm |
Translucent Orange | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now